Specifications
- Volume:500 ml
- Colour:Clear
- Description:Repositionable spray adhesive 75
- Pk:12
Specifications
About this item
3M™ repositionable spray adhesive 75 is a versatile, pressure sensitive, adhesive spray suitable for temporary holding or positioning of lightweight materials for display or prior to final assembly. It adheres in seconds, yet has an extra-long tack range that allows lifting and repositioning of materials.
- Hold and position lightweight materials temporarily
- Low soak-in formula allows it to bond delicate substrates without damaging the material
- Formula resists bleeding and staining for reliable and effective performance
- Dries clear, produces a smooth glue line, and resists wrinkling on most materials for a clean appearance
It is a clear adhesive which will not bleed through, stain, or wrinkle most materials. Neither will it soak through delicate substrates like foil, paper, and fabric. Suitable for bonding paper, tissue, cardboard, acetate film, foils, fabrics, plastic sheeting, or rubber to themselves, and to metal, wood, glass, and rigid plastics. It is widely used in textile cutting rooms for holding patterns in place instead of pinning them, and to prevent transfers moving during printing. It is also used for affixing removable labels.