Order Entry
Denmark
ContactUsLinkComponent
Calcium, Ion chromatography standard solution, Specpure®
Calcium, Ion chromatography standard solution, Specpure®
Catalog # 47234.AE
CAS Number:  
Calcium, Ion chromatography standard solution, Specpure®
Catalog # 47234.AE
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item