Order Entry
Denmark
ContactUsLinkComponent
Petroleum spirit, 40-60°C for gas chromatography, Burdick & Jackson™
Petroleum spirit, 40-60°C for gas chromatography, Burdick & Jackson™
Catalog # HONC65748-200L-RC
CAS Number:  
Petroleum spirit, 40-60°C for gas chromatography, Burdick & Jackson™
Catalog # HONC65748-200L-RC
CAS Number:  

Specifications

  • Pack type:
    Stainless steel drum, returnable
  • Pk:
    200 L
  • Seq:
    5549

Specifications

About this item

 :
Boiling Pt:40…60 °C
Flash Pt:–30 °C
UN:1268
ADR:3,II
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849