Order Entry
Denmark
ContactUsLinkComponent
Lead(II) acetate basic, anhydrous ≥33.0% basic Pb (PbO), ≥75.0% total Pb (PbO) ACS for the analysis of sugar acc. to horne, Fluka™
Environment
Irritant
Health
Lead(II) acetate basic, anhydrous ≥33.0% basic Pb (PbO), ≥75.0% total Pb (PbO) ACS for the analysis of sugar acc. to horne, Fluka™
Catalog # HONC32306H-1KG
CAS Number:  
Environment
Irritant
Health
Lead(II) acetate basic, anhydrous ≥33.0% basic Pb (PbO), ≥75.0% total Pb (PbO) ACS for the analysis of sugar acc. to horne, Fluka™
Catalog # HONC32306H-1KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Total Pb (as PbO):
    Min. 75.0 %
  • Basic Pb (as PbO):
    Min. 33.0 %
  • Loss on drying (105°C, 2 h):
    Max. 1.0 %
  • Water insoluble matter:
    Max. 1.0 %
  • Insoluble in acetic acid:
    Max. 0.02 %
  • Calcium (Ca):
    Max. 0.005 %
  • Cadmium (Cd):
    Max. 0.001 %
  • Cobalt (Co):
    Max. 0.001 %
  • Copper (Cu):
    Max. 0.002 %
  • Iron (Fe):
    Max. 0.002 %
  • Potassium (K):
    Max. 0.01 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.01 %
  • Nickel (Ni):
    Max. 0.001 %
  • Zinc (Zn):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.003 %
  • Nitrate (NO₃):
    Max. 0.003 %

Specification Test Results

About this item

 :
Density:3.58 g/cm3
UN:2291
ADR:6.1,III
 :807.72 g/mol
 :GHS09
 :GHS07
 :GHS08
CAS Number:51404-69-4
Formula:(CH₃COO)₂Pb·2Pb(OH)₂
MDL Number:MFCD00011154