Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Sodium azide

Plastic bottle, 1 kg
Catalog # HONC13412H-1KG
Environment
Health
Toxic
Supplier:   Honeywell Chemicals
Sodium azide
Catalog # HONC13412H-1KG
CAS Number:  26628-22-8
Sodium azide

Plastic bottle, 1 kg

CAS Number:  26628-22-8
Price
2.680,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol
  • Melting Pt:300 °C
  • Density:1.85 g/cm3 (20 °C)
  • Storage Temperature:Ambient
  • UN:1687
  • ADR:6.1,II
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Loss on dr. on P₂O₅, 20 mbar:
    Max. 0.2 %
  • Free alkali (as NaOH):
    Max. 0.2 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Water insoluble matter:
    Max. 0.05 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications