Cryogenic Vials, True North®
About this item
These externally-threaded vials with a flush cap design allow you to easily fill up all standard-sized cryogenic storage boxes including the 10×10 format.
- Silicone lid seal and patented easy-grip faceted lid with ergonomic 1,5 twist to open and close
- Recommended for mechanical freezers and vapor phase LN₂ (–196 °C)
- Puncture/rupture-resistance polypropylene vial
- Sterile, non-pyrogenic and non-cytotoxic
- Printed writing area and graduations
- Disposable
These True North® cryogenic vials can increase the freezers holding capacity by up to 23%.
Certifications: Includes traceable sterility certificate.
Ordering information: Assorted vials come in blue, red, green, purple and grey.
Packaging: Sterile packed in resealable bags.
2 Options Available Below
Product Details & Documents
These externally-threaded vials with a flush cap design allow you to easily fill up all standard-sized cryogenic storage boxes including the 10×10 format.
- Silicone lid seal and patented easy-grip faceted lid with ergonomic 1,5 twist to open and close
- Recommended for mechanical freezers and vapor phase LN₂ (–196 °C)
- Puncture/rupture-resistance polypropylene vial
- Sterile, non-pyrogenic and non-cytotoxic
- Printed writing area and graduations
- Disposable
These True North® cryogenic vials can increase the freezers holding capacity by up to 23%.
Certifications: Includes traceable sterility certificate.
Ordering information: Assorted vials come in blue, red, green, purple and grey.
Packaging: Sterile packed in resealable bags.



