Order Entry
Denmark
ContactUsLinkComponent
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Catalog # 33501-200KG
CAS Number:  
undefined
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Catalog # 33501-200KG
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    200 kg
  • Seq:
    4676

Specifications

Specification Test Results

  • Assay (GC):
    min. 99.5 %
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Ammonia (NH₃):
    Max. 0.002 %
  • Copper (Cu):
    Max. 0.0005 %
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.001 %
  • KMnO₄ reducing matter (as O):
    complying
  • Solubility in water:
    complying

Specification Test Results

About this item

 :
Boiling Pt:115 °C (1.013 hPa)
Melting Pt:-42 °C
Density:0.98 g/cm3 (20 °C)
Storage Temperature:Ambient
ADR:3,II
 :79.1 g/mol
UN:1282
 :GHS02
 :GHS07
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732
Melting Pt:–42 °C