Lead ionophore IV, Selectophore™ ionophore, Supelco®
Catalog # 15343-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:1053.59 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:145237-46-3
Formula:C₆₀H₈₄N₄O₄S₄
MDL Number:MFCD00679123