Order Entry
Denmark
ContactUsLinkComponent
Bis(3-methyl-1-phenyl-5-pyrazolone) ≥98.0% derivatisation reagent, for TLC, for cyanide determination, Supelco®
  15156-5G
 :  Merck
 :  
undefined
Bis(3-methyl-1-phenyl-5-pyrazolone) ≥98.0% derivatisation reagent, for TLC, for cyanide determination, Supelco®
  15156-5G
 :  Merck
 :  
  • Pk:
    5 g
  • Seq:
    3173
 :
 :346.39 g/mol
 :M-Clarity™ quality level = MQ100
 :7477-67-0
 :C₂₀H₁₈N₄O₂
 :MFCD00020765