Ordreindtastning
Denmark
ContactUsLinkComponent
Hydroxyethansyre ≥99.0% (by acidimetry), GPR RECTAPUR®
Corrosive
Irritant
Hydroxyethansyre ≥99.0% (by acidimetry), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Irritant
Hydroxyethansyre ≥99.0% (by acidimetry), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    White to off-white crystals
  • Assay (acidimetric):
    min. 99.0 %
  • Water:
    max. 0.5 %
  • Melting range (lower value):
    min. 75 °C
  • Melting range (upper value):
    max. 79 °C
  • Identity (IR):
    Passes test

Specification Test Results

Frequently Bought Together