Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1-Methylnicotinamide chloride
Supelco® Organic Reference Standard, 1-Methylnicotinamide chloride
Catalog # M4627-1G
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 1-Methylnicotinamide chloride
Catalog # M4627-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Standards:
    Analytical standard
  • CAS No.:
    1005-24-9
  • Description:
    1-Methylnicotinamide chloride
  • Storage conditions:
    RT
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00060042
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N-Methylnicotinic acid amide chloride
  • Formula:
    C₇H₉N₂OCl
  • Molecular weight:
    172,61

Specifications

About this item