Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, Vitamin B₁₂ₐ hydrochloride
Supelco® Organic Reference Standard, Vitamin B₁₂ₐ hydrochloride
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Vitamin B₁₂ₐ hydrochloride
Supplier:  Merck

Specifications

  • CAS No.:
    59461-30-2
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00167463
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Hydroxocobalamin hydrochloride
  • Molecular weight:
    1382,82

Specifications

Frequently Bought Together