Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, 4-Deoxypyridoxine hydrochloride
Supelco® Organic Reference Standard, 4-Deoxypyridoxine hydrochloride
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, 4-Deoxypyridoxine hydrochloride
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    148-51-6
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00051296
  • M-Clarity quality level:
    MQ100
  • Melting point:
    263 - 265 °C
  • Molecular weight:
    189,64

Specifications

Related Information

Frequently Bought Together