0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Standards
Organic Standards
Levetiracetam, analytical standard, Supelco®
Levetiracetam, analytical standard, Supelco®
Catalog # 78645-10MG
Supplier:
Merck
CAS Number:
Expand image
Levetiracetam, analytical standard, Supelco®
Catalog # 78645-10MG
Supplier:
Merck
CAS Number:
Specifications
Pk:
10 mg
Standards:
analytical standard
CAS No.:
102767-28-2
Description:
Levetiracetam
Description:
Levetiracetam
Specifications
About this item
Product Details
Product Details