Formamide solution, NMR reference standard, Supelco®
Supplier: Merck
About this item
3 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 551384-8IN
- 611891-1EA
- 736201-8IN
- Description:
- Formamide solution
- Formamide solution
- Formamide solution
Specifications
Product Family Options
Product Information
- ConcentrationStandardsPk InfoPkAvailabilityPrice
- 551384-8INFormamide solutionFormamide solution90% in DMSO-d6 (99.9 atom % D)NMR reference standardNMR tube size 5 mm x 8 in.1Log in for availability1 * 1 Each1.730,00 krSpecifications:
More
Specifications
Cat. No.551384-8INDescriptionFormamide solution - 611891-1EAFormamide solutionFormamide solution45% in DMSO-d6 (99.9 atom % D)NMR reference standardNMR tube size 5 mm x 8 in.1Log in for availability1 * 1 Each2.690,00 krSpecifications:
More
Specifications
Cat. No.611891-1EADescriptionFormamide solution - 736201-8INFormamide solutionFormamide solution90% in DMSO-d6 (99.9 atom % D)NMR reference standardNMR tube size 5 mm x 8 in.1Log in for availability1 * 1 Items4.970,00 krSpecifications:
More
Specifications
Cat. No.736201-8INDescriptionFormamide solution