Order Entry
Denmark
ContactUsLinkComponent
Zearalanone solution, analytical standard, Supelco®
Zearalanone solution, analytical standard, Supelco®
Catalog # 32193-1.2ML
Supplier:  Merck
CAS Number:  
undefined
Zearalanone solution, analytical standard, Supelco®
Catalog # 32193-1.2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 mug/mL in acetonitrile
  • Pk:
    1,2 mL
  • Standards:
    analytical standard
  • CAS No.:
    5975-78-0
  • Description:
    Zearalanone solution
  • Description:
    Zearalanone solution

Specifications

About this item