Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, N-Acetyl-S-Phenyl-L-Cysteine
Supelco® Organic Reference Standard, N-Acetyl-S-Phenyl-L-Cysteine
Catalog # 30498-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, N-Acetyl-S-Phenyl-L-Cysteine
Catalog # 30498-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    4775-80-8
  • Description:
    N-Acetyl-S-phenyl-L-cysteine
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD16661257
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    239,29
  • Flash point:
    Not applicable

Specifications

About this item