Order Entry
Denmark
ContactUsLinkComponent
Iron hematoxylin kit (Weigert's) stain for histology, Sigma-Aldrich®
Iron hematoxylin kit (Weigert's) stain for histology, Sigma-Aldrich®
Catalog # 1.15973.0002
Supplier:  Merck
CAS Number:  
undefined
Iron hematoxylin kit (Weigert's) stain for histology, Sigma-Aldrich®
Catalog # 1.15973.0002
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    2
  • Seq:
    0002
  • Pk Info:
    2×500 ml (Preparation Capacity 400 - 500 slides)

Specifications

About this item

 :
Storage Temperature:Ambient
UN:1170
ADR:3,
 :GHS02
 :GHS07
 :1
MDL Number:MFCD00284879