Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Tyramine, analytical standard, Supelco®
Organic reference standard, Tyramine, analytical standard, Supelco®
Catalog # 80345-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Tyramine, analytical standard, Supelco®
Catalog # 80345-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    51-67-2
  • Description:
    Tyramine, ≥98,5% (GC)
  • Melting point:
    160...162 °C (lit.)
  • Formula:
    C₈H₁₁NO
  • Storage temperature:
    2...8 °C
  • Description:
    Tyramine
  • Boiling point:
    175...181 °C/8 mm Hg (lit.)
  • Form:
    Neat

Specifications

About this item

Can enter catecholaminergic terminals and be released as a false transmitter.