Supelco® Organic Reference Standard, Trigonelline hydrochloride
61407-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:6138-41-6
- Description:Trigonelline hydrochloride
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00077250
- M-Clarity quality level:MQ100
- Synonym(s):1-Methylpyridinium-3-carboxylate hydrochloride; N-Methylnicotinic acid betaine hydrochloride
- Molecular weight:173,60