Organic reference standard, Triethylamine, analytical standard, Supelco®
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:121-44-8
- Melting point:−115 °C
- Formula:C₆H₁₅N
- Description:Triethylamine
- Flash point:–15 °C
Specifications
Frequently Bought Together