Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Triacetin, analytical standard, Supelco®
Organic reference standard, Triacetin, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, Triacetin, analytical standard, Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    102-76-1
  • Melting point:
    3 °C
  • Formula:
    C₉H₁₄O₆
  • Description:
    Triacetin
  • Flash point:
    138 °C

Specifications

Frequently Bought Together