Supelco® Organic Reference Standard, trans-β-Farnesene
73492-1ML-F
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:18794-84-8
- Description:trans-β-Farnesene
- Storage conditions:–20 °C
- Hazard class description:H304
- Shipping conditions:WICE
- MDL No.:MFCD00065433
- M-Clarity quality level:MQ100
- Synonym(s):trans-7,11-Dimethyl-3-methylene-1,6,10-dodecatriene
- Molecular weight:204,35
- Flash point:110 °C
- GHS signal: GHS07
: P301 + P310 + P331