Organic reference standard, trans-2-Nonenal, analytical standard, Supelco®
07592-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:18829-56-6
- Description:trans-2-Nonenal, ≥95,0% (GC)
- Formula:C₉H₁₆O
- Description:trans-2-Nonenal
- Boiling point:88...90 °C/12 mm Hg (lit.)