Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, tert-Butylhydroquinone, analytical standard, Supelco®
Organic reference standard, tert-Butylhydroquinone, analytical standard, Supelco®
Catalog # 07293-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, tert-Butylhydroquinone, analytical standard, Supelco®
Catalog # 07293-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1948-33-0
  • Description:
    tert-Butylhydroquinone, ≥98,0% (GC)
  • Melting point:
    127...129 °C
  • Formula:
    C₁₀H₁₄O₂
  • Description:
    tert-Butylhydroquinone

Specifications

About this item