Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, Terpinolene, analytical standard, Supelco®
  43905-1ML
 :  Merck
 :  
undefined
Organic reference standard, Terpinolene, analytical standard, Supelco®
  43905-1ML
 :  Merck
 :  

 

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    586-62-9
  • Description:
    Terpinolene, ≥94% (GC)
  • Melting point:
    184...185 °C(lit.)
  • Formula:
    C₇H₈N₄O₂
  • Storage temperature:
    −20°C
  • Description:
    Terpinolene
  • Form:
    Neat