Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Terpineol, mixture of isomers, analytical standard, Supelco®
Organic reference standard, Terpineol, mixture of isomers, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, Terpineol, mixture of isomers, analytical standard, Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    8000-41-7
  • Formula:
    C₁₀H₁₈O
  • Description:
    Terpineol
  • Boiling point:
    213...218 °C(lit.)

Specifications

Frequently Bought Together