Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Tartrazine, analytical standard, Supelco®
Organic reference standard, Tartrazine, analytical standard, Supelco®
Catalog # 03322-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Tartrazine, analytical standard, Supelco®
Catalog # 03322-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1934-21-0
  • Description:
    Tartrazine, ≥99,0% (HPLC)
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₆H₉N₄Na₃O₉S₂
  • Description:
    Tartrazine

Specifications

About this item