Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Sudan I-d5, analytical standard, Supelco®
Organic reference standard, Sudan I-d5, analytical standard, Supelco®
Catalog # 34184-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Sudan I-d5, analytical standard, Supelco®
Catalog # 34184-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    752211-63-5
  • Description:
    Sudan I-d5
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₂D₆H₁₀N₄O
  • Description:
    Sudan I-d5
  • Form:
    Neat

Specifications

About this item