Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, S-Methyl methanethiosulfonate, analytical standard, Supelco®
Organic reference standard, S-Methyl methanethiosulfonate, analytical standard, Supelco®
Catalog # 49983-250MG
Supplier:  Merck
CAS Number:  
Organic reference standard, S-Methyl methanethiosulfonate, analytical standard, Supelco®
Catalog # 49983-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2949-92-0
  • Description:
    S-Methyl methanethiosulfonate
  • Formula:
    C₂H₆O₂S₂
  • Storage temperature:
    2...8 °C
  • Description:
    S-Methyl methanethiosulfonate
  • Boiling point:
    69...71 °C/0.4 mmHg

Specifications

About this item