Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Rhodamine 6G, analytical standard, Supelco®
Organic reference standard, Rhodamine 6G, analytical standard, Supelco®
Catalog # 56226-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Rhodamine 6G, analytical standard, Supelco®
Catalog # 56226-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    989-38-8
  • Description:
    Rhodamine 6G
  • Formula:
     C₂₈H₃₁N₂O₃Cl
  • Description:
    Rhodamine 6G
  • Form:
    Neat

Specifications

About this item

A rhodamine analog useful in Pgp efflux assays. Has been used to characterize kinetics of MRP1- mediated efflux.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.