Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Rebaudioside B, analytical standard, Supelco®
Organic reference standard, Rebaudioside B, analytical standard, Supelco®
Catalog # 49747-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Rebaudioside B, analytical standard, Supelco®
Catalog # 49747-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    58543-17-2
  • Description:
    Rebaudioside B, ≥95,0% (HPLC)
  • Formula:
    C₃₈H₆₀O₁₈
  • Description:
    Rebaudioside B
  • Form:
    Neat

Specifications

About this item