Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, p-Tolualdehyde, analytical standard, Supelco®
Organic reference standard, p-Tolualdehyde, analytical standard, Supelco®
  41423-5ML
 :  Merck
 :  
undefined
Organic reference standard, p-Tolualdehyde, analytical standard, Supelco®
  41423-5ML
 :  Merck
 :  

 

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    104-87-0
  • Description:
    p-Tolualdehyde, ≥97,0% (GC)
  • Formula:
    C₈H₈O
  • Description:
    p-Tolualdehyde
  • Flash point:
    72 °C
  • Boiling point:
    204…205 °C
  • Form:
    Neat