Organic reference standard, Propylamine, analytical standard, Supelco®
53646-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:107-10-8
- Description:Propylamine, ≥99,0% (GC)
- Melting point:–83 °C
- Formula:C₃H₉N
- Description:Propylamine
- Flash point:–30 °C
- Boiling point:48 °C
- Form:Neat