Supelco® Organic Reference Standard, Physcion
Catalog # 93893-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:521-61-9
- Description:Physcion
- Storage conditions:2…8 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00017374
- M-Clarity quality level:MQ100
- Synonym(s):1,8-Dihydroxy-3-methoxy-6-methylanthraquinone; 6-O-Methylemodin; 6-Methoxychrysophanic acid; Emodin-3-methyl ether; Parietin; Rheochrysidin
- Molecular weight:284,26
Specifications
About this item
- GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501