Organic reference standard, Phthalide, analytical standard, Supelco®
Catalog # 55906-100MG
Supplier: Merck
CAS Number:
Specifications
- Pakn:100 mg
- Standard:Analytical standard
- CAS-nr:87-41-2
- Beskrivelse:Phthalide, ≥99,0% (GC)
- Melting point:71...74 °C
- Formel:C₈H₆O₂
- Beskrivelse:Phthalide
- Flammepunkt:152 °C
- Kogepunkt:290 °C
- Form:Neat