Organic reference standard, Patulin-13C7 solution, analytical standard, Supelco®
Catalog # 35516-1.2ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:25 µg/ml in acetonitrile
- Pk:1,2 mL
- Standards:Analytical standard
- CAS No.:1353867-99-8
- Description:Patulin-13C7 solution
- Formula:₁₃C₇H₆O₄
- Storage temperature:–20 °C
- Description:Patulin-13C7 solution
- Flash point:2 °C