Organic reference standard, Oxybenzone, analytical standard, Supelco®
Oxybenzone, ≥98% (GC), Analytical standard, Neat, 50 mg
Catalog # 59647-50MG
Supplier: Merck
Organic reference standard, Oxybenzone, analytical standard, Supelco®
Catalog # 59647-50MG
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Specifications
Specifications
- Pk:50 mg
- Standards:Analytical standard
- Description:Oxybenzone, ≥98% (GC)
- Form:Neat
- Cat. No.:59647-50MG
- Formula:C₁₄H₁₂O₃
- Description:Oxybenzone
- Boiling point:150...160 °C
- Storage temperature:2...8 °C
- Flash point:100 °C
- Melting point:62...64 °C
- CAS No.:131-57-7



