Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Oil Orange SS, analytical standard, Supelco®
Organic reference standard, Oil Orange SS, analytical standard, Supelco®
  79285-25MG
 :  Merck
 :  
undefined
Organic reference standard, Oil Orange SS, analytical standard, Supelco®
  79285-25MG
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2646-17-5
  • Description:
    Oil Orange SS
  • Melting point:
    124...126 °C
  • Formula:
    C₁₇H₁₄N₂O
  • Description:
    Oil Orange SS
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.