Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, Octyl acetate, analytical standard, Supelco®
Catalog # 04622-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Octyl acetate, analytical standard, Supelco®
Catalog # 04622-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    112-14-1
  • Description:
    Octyl acetate, ≥98,5% (GC)
  • Formula:
    C₁₀H₂₀O₂
  • Description:
    Octyl acetate
  • Boiling point:
    211 °C

Specifications

About this item