Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Ochratoxin A-d5 solution, analytical standard, Supelco®
Organic reference standard, Ochratoxin A-d5 solution, analytical standard, Supelco®
Catalog # 32245-2ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Ochratoxin A-d5 solution, analytical standard, Supelco®
Catalog # 32245-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in acetonitrile
  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • CAS No.:
    666236-28-8
  • Description:
    Ochratoxin A-d5 solution
  • Formula:
    C₂₀D₅H₁₃ClNO₆
  • Storage temperature:
    –20 °C
  • Description:
    Ochratoxin A-d5 solution

Specifications

About this item