Organic reference standard, Ochratoxin A solution, analytical standard, Supelco®
34037-2ML-R
: Merck
:
- Concentration:10 µg/ml in acetonitrile
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:303-47-9
- Description:Ochratoxin A solution
- M-Clarity quality level:MQ100
- Formula:C₂₀H₁₈ClNO₆
- Storage temperature:–20 °C
- Description:Ochratoxin A solution