Organic reference standard, Nivalenol hydrate, analytical standard, Supelco®
Catalog # 32929-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:23282-20-4
- Description:Nivalenol hydrate
- M-Clarity quality level:MQ100
- Formula:C₁₅H₂₀O₇
- Storage temperature:−20 °C
- Description:Nivalenol hydrate
- Form:Neat