Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Neoxanthin, analytical standard, Supelco®
Organic reference standard, Neoxanthin, analytical standard, Supelco®
Catalog # 72994-1MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Neoxanthin, analytical standard, Supelco®
Catalog # 72994-1MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Standards:
    Analytical standard
  • CAS No.:
    14660-91-4
  • Description:
    Neoxanthin, ≥97,0% (HPLC)
  • Formula:
    C₄₀H₅₆O₄
  • Storage temperature:
    –20 °C
  • Description:
    Neoxanthin

Specifications

About this item