Organic reference standard, Methyl cyclo pentenolone, analytical standard, Supelco®
Catalog # 80133-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:765-70-8
- Description:Methyl cyclopentenolone, ≥98,0% (GC)
- Formula:C₆H₈O₂
- Description:Methyl cyclopentenolone
- Boiling point:104…108 °C
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.