Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Maltol, analytical standard, Supelco®
Organic reference standard, Maltol, analytical standard, Supelco®
Catalog # 18299-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Maltol, analytical standard, Supelco®
Catalog # 18299-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    118-71-8
  • Description:
    Maltol, ≥97,0% (GC)
  • Melting point:
    160…164 °C
  • Formula:
    C₆H₆O₃
  • Description:
    Maltol
  • Form:
    Neat

Specifications

About this item