Supelco® Organic Reference Standard, Luteolin 7-glucoside
49968-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:5373-11-5
- Description:Luteolin 7-glucoside
- Storage conditions:2…8 °C
- Shipping conditions:WICE
- MDL No.:MFCD06799436
- M-Clarity quality level:MQ100
- Synonym(s):Luteolin 7-O-β-ᴅ-glucoside; 3′,4′,5,7-Tetrahydroxyflavone 7-glucoside; Cynaroside; Glucoluteolin; Glucosylluteolin; Luteoloside
- Molecular weight:448,38