Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, Linalyl acetate, analytical standard, Supelco®
Catalog # 49599-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Linalyl acetate, analytical standard, Supelco®
Catalog # 49599-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    115-95-7
  • Description:
    Linalyl acetate, ≥97,0% (GC)
  • Formula:
    C₁₂H₂₀O₂
  • Storage temperature:
    2…8 °C
  • Description:
    Linalyl acetate
  • Boiling point:
    220 °C
  • Form:
    Neat

Specifications

About this item