Organic reference standard, Leucomalachite Green-d5, analytical standard, Supelco®
Catalog # 34182-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:947601-82-3
- Description:Leucomalachite Green-d5
- M-Clarity quality level:MQ100
- Formula:C₂₃H₂₁D₅N₂
- Description:Leucomalachite Green-d5
- Form:Neat