Organic reference standard, Leucomalachite Green, analytical standard, Supelco®
69669-25MG
: Merck
:
- Pakn:25 mg
- Standard:Analytical standard
- CAS-nr:129-73-7
- Beskrivelse:Leucomalachite Green, ≥98,0% (HPLC)
- M-Clarity quality level:MQ100
- Melting point:100…102 °C
- Formel:C₂₃H₂₆N₂
- Beskrivelse:Leucomalachite Green
- Form:Neat
: Bottomless glass bottle. Contents are inside inserted fused cone.